Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Strontium carbonate ≥99.9% (Sr basis) (low alkali and heavy metals)


93-3803-50G 93-3803-250G
100211-254EA 104.74 USD InStock
100211-254 100202-452
Strontium carbonate ≥99.9% (Sr basis) (low alkali and heavy metals)
Strontium carbonate


Formula: CO₃Sr
MW: 147.63
Boiling Pt: 2647
Melting Pt: 1768.85
Density: 3.70
Storage Temperature: Ambient
MDL Number: MFCD00011250
CAS Number: 1633-05-2
EINECS: 216-643-7
Merck Index: 14,08838
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty