Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diyttrium trioxide ≥99.9% (Y basis)

Warning

93-3911-100G 93-3911-25G 93-3911-500G
100207-046EA 94.83 USD InStock
100207-046 100210-390 100200-138
Diyttrium trioxide ≥99.9% (Y basis)
Diyttrium trioxide


Formula: Y₂O₃
Boiling Pt: 4300 °C (1013 hPa)
Melting Pt: 2410 °C
Density: 5.01 g/cm³ (20 °C)
MDL Number: MFCD00011473
CAS Number: 1314-36-9
EINECS: 215-233-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty