Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diyttrium trioxide ≥99.999% (Y basis), PURATREM

Warning

93-3925-10G 93-3925-1000G 93-3925-50G
100200-560EA 73.6 USD InStock
100200-560 100201-278 100198-376
Diyttrium trioxide ≥99.999% (Y basis), PURATREM
Diyttrium trioxide


Formula: Y₂O₃
Boiling Pt: 4300 °C (1013 hPa)
Melting Pt: 2410 °C
Density: 5.01 g/cm³ (20 °C)
MDL Number: MFCD00011473
CAS Number: 1314-36-9
EINECS: 215-233-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty