Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Silver(II) fluoride ≥98%

Danger

93-4747-25G 93-4747-5G
100206-152EA 423.2 USD InStock
100206-152 100204-338
Silver(II) fluoride ≥98%
Silver(II) fluoride


Formula: AgF₂
MW: 145.86 g/mol
Boiling Pt: 700 °C (dec.)
Melting Pt: 690
Density: 4.58
MDL Number: MFCD00003411
CAS Number: 7783-95-1
EINECS: 232-037-5
UN: 1759
ADR: 8 III
Merck Index: 12,08656
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty