Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tin(II) fluoride ≥97.5%

Danger

93-5033-250G 93-5033-50G
100209-142EA 176.92 USD InStock
100209-142 100207-858
Tin(II) fluoride ≥97.5%
Tin(II) fluoride


Formula: SnF₂
MW: 156.71
Boiling Pt: 850
Melting Pt: 215
Density: 4.57 g/cm³ (20 °C)
MDL Number: MFCD00042540
CAS Number: 7783-47-3
EINECS: 231-999-3
UN: 1759
ADR: 8 II
Merck Index: 12,08940
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty