Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(tributyltin)sulphate ≥97%

Warning

93-5058-100G 93-5058-25G
100205-626EA 171.26 USD InStock
100205-626 100196-714
Bis(tributyltin)sulphate ≥97%
Bis(tributyltin)sulphate


Formula: C₂₄H₅₄O₄SSn₂
MW: 676.18
MDL Number: MFCD00185908
CAS Number: 26377-04-8
EINECS: 247-634-6
UN: 3146
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty