Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cesium hydroxide hydrate ≥99.9% (Cs basis)

Danger

93-5521-10G 93-5521-50G
100210-476EA 55.2 USD InStock
100210-476 100210-478
Cesium hydroxide hydrate ≥99.9% (Cs basis)
Cesium hydroxide hydrate


Formula: HCsO·H₂O
Melting Pt: 272 °C
Density: 3.675
MDL Number: MFCD00010964
CAS Number: 12260-45-6
EINECS: 244-344-1
UN: 2682
ADR: 8,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty