Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Praseodymium ≥99.9%, ingot

Warning

93-5942-50G 93-5942-10G
100200-778EA 961.05 USD InStock
100200-778 100200-776
Praseodymium ≥99.9%, ingot
Praseodymium


Formula: Pr
MW: 140.91
Boiling Pt: 3127 °C
Melting Pt: 935 °C
Density: 6.782
MDL Number: MFCD00011174
CAS Number: 7440-10-0
EINECS: 231-120-3
UN: 2813
ADR: 4.3 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty