Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Samarium(III) oxide ≥99.9% (Sm basis)


93-6211-25G 93-6211-500G 93-6211-100G
100196-794EA 108.98 USD InStock
100196-794 100201-136 100199-646
Samarium(III) oxide ≥99.9% (Sm basis)
Samarium(III) oxide


Formula: O₃Sm₂
MW: 348.72 g/mol
Melting Pt: 2325
Density: 8.347
Storage Temperature: Ambient
MDL Number: MFCD00011237
CAS Number: 12060-58-1
EINECS: 235-043-6
Merck Index: 14,08347
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty