Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Digadolinium trioxide ≥99.99% (Gd basis), PURATREM

Warning

93-6406-500G 93-6406-100G
100197-260EA 806.77 USD InStock
100197-260 100207-132
Digadolinium trioxide ≥99.99% (Gd basis), PURATREM
Digadolinium trioxide


Formula: Gd₂O₃
MW: 362.5 g/mol
Melting Pt: 2410
Density: 7.407
MDL Number: MFCD00011026
CAS Number: 12064-62-9
EINECS: 235-060-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty