Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dysprosium ≥99.9%, ingot


93-6637-100G 93-6637-25G
100196-994EA 717.6 USD InStock
100196-994 100209-448
Dysprosium ≥99.9%, ingot
Dysprosium


Formula: Dy
MW: 162.5
Boiling Pt: 2335 °C
Melting Pt: 1409 °C
Density: 8.54
MDL Number: MFCD00010982
CAS Number: 7429-91-6
EINECS: 231-073-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty