Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Holmium trinitrate hydrate ≥99.9% (Ho basis)

Danger

93-6712-25G 93-6712-5G
100203-560EA 251.94 USD InStock
100203-560 100210-530
Holmium trinitrate hydrate ≥99.9% (Ho basis)
Holmium trinitrate hydrate


Formula: HoN₃O₉·5H₂O
Melting Pt: 83.74 - 84.62 °C
MDL Number: MFCD00149737
CAS Number: 14483-18-2
EINECS: 233-438-8
UN: 1477
ADR: 5.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty