Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hafnium dioxide ≥98%


93-7204-50G 93-7204-10G
100203-578EA 257.6 USD InStock
100203-578 100204-420
Hafnium dioxide ≥98%
Hafnium dioxide


Formula: HfO₂
Boiling Pt: 5400 °C (1013 hPa)
Melting Pt: 2812 °C
Density: 9.68 g/cm³ (20 °C)
MDL Number: MFCD00003565
CAS Number: 12055-23-1
EINECS: 235-013-2
Merck Index: 14,04727
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty