Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tantalum(V) oxide ≥99.8% (99.99+%-Ta), PURATREM


93-7306-50G 93-7306-10G
100202-286EA 297.23 USD InStock
100202-286 100202-284
Tantalum(V) oxide ≥99.8% (99.99+%-Ta), PURATREM
Tantalum(V) oxide


Formula: O₅Ta₂
Melting Pt: 1800 °C
Density: 8.2 g/cm³ (20 °C)
MDL Number: MFCD00011254
CAS Number: 1314-61-0
EINECS: 215-238-2
Merck Index: 14,09056
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty