Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thallium(I) iodide ≥99.999% (Tl basis), PURATREM

Danger

93-8108-25G 93-8108-5G
100203-594EA 443.02 USD InStock
100203-594 100205-782
Thallium(I) iodide ≥99.999% (Tl basis), PURATREM
Thallium(I) iodide


Formula: ITl
MW: 331.29 g/mol
Boiling Pt: 823
Melting Pt: 440
Density: 7.09
MDL Number: MFCD00011279
CAS Number: 7790-30-9
EINECS: 232-199-7
UN: 1707
ADR: 6.1 II
Merck Index: 13,09335
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty