Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Acetylnaphthalene suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

A2080-25GM A2080-100GM
700004-020EA 76.7 USD InStock
700004-020 700004-018
2-Acetylnaphthalene suitable for use as excipient
2-Acetylnaphthalene
2'-Acetonaphthone , also know as Methyl beta-naphthyl ketone, is used in anti-odor applications such as air purifiers.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 52…54 °C
Density: 1.1171 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004108
CAS Number: 93-08-3
EINECS: 202-216-2
UN: 3077
ADR: 9 III
Order Now
Specification Test Results
Assay 98.5% Min.

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty