Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyltriethylammonium chloride for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

B1083-100GM B1083-500GM
700007-534EA 125.5 USD InStock
700007-534 700007-538
Benzyltriethylammonium chloride for synthesis
Benzyltriethylammonium chloride
Benzyltriethylammonium Chloride, also known as Triton B, is a quaternary ammonium salt that functions as an organic base and is a popular phase-transfer catalyst.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₁₃H₂₂ClN
MW: 227.78 g/mol
Melting Pt: 190…192 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011824
CAS Number: 56-37-1
EINECS: 200-270-1
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty