Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(2-methoxyethyl)ether ≥99%, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

B1099-200LTB B1099-250ML B1099-4LTPV
700003-402EA 9027.45 USD InStock
700003-402 700003-404 700003-406
Bis(2-methoxyethyl)ether ≥99%, Reagent Grade
Bis(2-methoxyethyl)ether
Bis(2-methoxyethyl) Ether, Reagent is more commonly referred to as Diglyme and is an organic compound that is a clear and colorless liquid with a faint ether like smell. It is solvent that has a high boiling point and that is miscible with water, alcohol as well as hydrocarbon solvents. Its main use is as a solvent for organic reactions as well as being able to chelate small cations. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.


Formula: C₆H₁₄O₃
Boiling Pt: 162 °C (1013 hPa)
Melting Pt: –64 °C
Density: 0.944 g/cm³ (20 °C)
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00008503
CAS Number: 111-96-6
EINECS: 203-924-4
UN: 3271
ADR: 3,III
Merck Index: 13,03187
Order Now
Specification Test Results
Assay 99% Min.

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty