Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl phthalate ≥98% NF suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

D1097-200LTB D1097-20LT D1097-4LTGL D1097-500MLG D1097-500MLG
700006-000EA 12028.05 USD InStock
700006-000 700006-002 95032-898 95032-900 700006-004
Diethyl phthalate ≥98% NF suitable for use as excipient
Diethyl phthalate
Diethyl Phthalate, NF - Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


Formula: C₁₂H₁₄O₄
MW: 222.24 g/mol
Boiling Pt: 298
Melting Pt: -3
Density: 1.121
Flash Pt: 156 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00009111
CAS Number: 84-66-2
EINECS: 201-550-6
Merck Index: 12,07527
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d0a6c6b14ead75148c0661141b5e8d00e6fc1a9&vid=3d0a6c6b14ead75148c0661141b5e8d00e6fc1a9&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 98% Min.

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty