Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Docusate sodium ≥99% USP suitable for use as excipient

Warning

DO105-2.5KG DO105-125GM
95033-012EA 2885.99 USD InStock
95033-012 700005-896 95033-010
Docusate sodium ≥99% USP suitable for use as excipient
Docusate sodium


Formula: C₂₀H₃₇NaO₇S
MW: 444.57 g/mol
Boiling Pt: ∼180 °C (1013 hPa)
Melting Pt: 155
Density: ∼1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012455
CAS Number: 577-11-7
EINECS: 209-406-4
Merck Index: 12,03460
Order Now
Specification Test Results
Assay 99% Min.

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty