Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

EDTA trisodium salt ≥99% for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP

E1046-125GM E1046-3KG E1046-500GM
700007-286EA 120.83 USD InStock
700007-286 700007-288 700007-292
EDTA trisodium salt ≥99% for synthesis
EDTA trisodium salt
Ethylenediaminetetraacetic Acid Trisodium Salt is a colorless aminopolycarboxylic acid that is water soluble. It is typically used to dissolve lime scale. It is valuable because of its role as a six toothed ligand and chelating agent. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.


Formula: C₁₀H₁₃N₂Na₃O₈
MW: 358.19 g/mol
Melting Pt: > 300 °C
Density: 0.7215 g/cm³ (20 °C)
Storage Temperature: Ambient
CAS Number: 150-38-9
EINECS: 205-758-8
Merck Index: 12,03558
Order Now
Specification Test Results
Assay 99% Min.

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty