Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl nicotinate ≥99% for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP

E2510-500GM E2510-100GM
700008-732EA 229.4 USD InStock
700008-732 700008-730
Ethyl nicotinate ≥99% for synthesis
Ethyl nicotinate
Ethyl Nicotinate is a water soluble, strongly basic compound.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 223
Density: 1.107
Storage Temperature: Ambient
MDL Number: MFCD00006389
CAS Number: 614-18-6
EINECS: 210-370-7
Order Now
Specification Test Results
Assay 99% Min.

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty