Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxybenzoic acid ≥99%, Reagent Grade for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

H1083-5KG H1083-500GM H1083-25KGBL H1083-100GM
700006-488EA 481.6 USD InStock
700006-488 700006-486 700006-484 700006-482
4-Hydroxybenzoic acid ≥99%, Reagent Grade for synthesis
4-Hydroxybenzoic acid
p-Hydroxybenzoic Acid, Reagent is also known as 4-Hydroxybenzoic acid and para-Hydroxybenzoic acid. It is mostly known as the basis for the preparation of its esters which are parabens. This is used in ophthalmic solutions and as a preservative in certain cosmetics. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.


Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 336
Melting Pt: 210…211 °C
Density: 1.398 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002547
CAS Number: 99-96-7
EINECS: 202-804-9
Merck Index: 13,04837
Order Now
Specification Test Results
Assay 99% Min.

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty