Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L(+)-Lysine monohydrate ≥98.5% suitable for use as excipient, for biochemistry

Supplier: SPECTRUM CHEMICAL MFG CORP

L1146-100GM L1146-1KGBL L1146-25GM L1146-5KGBL
700007-164EA 339.2 USD InStock
700007-164 700007-166 700007-168 700007-170
L(+)-Lysine monohydrate ≥98.5% suitable for use as excipient, for biochemistry
L(+)-Lysine monohydrate
Allergen statement (available upon request)


Formula: H₂N(CH₂)₄CH(NH₂)CO₂H·H₂O
MW: 164.2 g/mol
Melting Pt: 225
Storage Temperature: Refrigerator
MDL Number: MFCD00151035
CAS Number: 39665-12-8
EINECS: 200-294-2
Order Now
Specification Test Results
Assay 98.5% Min.

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty