Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Magnesium acetate tetrahydrate ≥98%, crystals ACS suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

M1020-12KG M1020-500GM M1020-2.5KG M1020-125GM
700004-342EA 2115.1 USD InStock
700004-342 700004-346 700004-344 700004-340
Magnesium acetate tetrahydrate ≥98%, crystals ACS suitable for use as excipient
Magnesium acetate tetrahydrate
Form: Crystal


Formula: (H₃CCOO)₂Mg·4H₂O
Melting Pt: 80 °C
Density: 1.454 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149214
CAS Number: 16674-78-5
EINECS: 205-554-9
Merck Index: 13,05676
Order Now
Specification Test Results
Assay 98% Min.

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty