Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium benzoate ≥99% NF

Supplier: SPECTRUM CHEMICAL MFG CORP

P1181-2.5KG P1181-12KG P1181-500GM
95035-088EA 347.9 USD InStock
95035-088 700005-612 95035-090
Potassium benzoate ≥99% NF
Potassium benzoate
Potassium Benzoate, NF is used as a preservative that limits the growth of yeast, mold and some bacteria. 

All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 


Formula: C₇H₅KO₂
Storage Temperature: Ambient
MDL Number: MFCD00013061
CAS Number: 582-25-2
EINECS: 209-481-3
Order Now
Specification Test Results
Assay 99% Min.

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty