Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phenol liquified ≥89% USP suitable for use as excipient

Danger

95031-342CS 3510.84 USD InStock
95031-342 95031-346 89078-924 700003-188 95031-344
Phenol liquified ≥89% USP suitable for use as excipient
Phenol liquified
Liquefied Phenol, USP is commonly used for permanent treatment of ingrown toe and finger nails, a procedure known as a chemical matrixectomy. All Spectrum Chemical USP products USP are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


Formula: C₆H₅OH
Density: 1.071
Storage Temperature: Ambient
MDL Number: MFCD00002143
CAS Number: 108-95-2
EINECS: 203-632-7
UN: 2821
ADR: 6.1,II
Order Now
Specification Test Results
Assay 89% Min.

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty