Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Polyvinylpyrrolidone, M.W. ∼ 10,000 K-15

Supplier: SPECTRUM CHEMICAL MFG CORP

PO146-5KG PO146-500GM PO146-100GM
700008-882EA 1735.25 USD InStock
700008-882 700008-880 700008-878
Polyvinylpyrrolidone, M.W. ∼ 10,000 K-15
Polyvinylpyrrolidone
Polyvinylpyrrolidone K-15 is also known as polyvidone or povidone. It has been used for trauma victims as a plasma volume expander, as well as a binder in some pharmaceutical tablets.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty