Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-(+)-Trehalose dihydrate ACS for biochemistry

Supplier: SPECTRUM CHEMICAL MFG CORP

T1097-50KGBL T1097-100GM T1097-1KGBL T1097-5KGBL T1097-25GM T1097-500GM T1097-5GM
700009-756EA 14414.6 USD InStock
700009-756 700009-748 700009-750 700009-760 700009-752 700009-754 700009-758
D-(+)-Trehalose dihydrate ACS for biochemistry
D-(+)-Trehalose dihydrate
D-(+)-Trehalose, Dihydrate, is a natural alpha-linked disaccharide that is also referred to as mycose or tremalose. It is found naturally in plants and microorganisms such as grasshoppers, shrimp, butterflies and bees. It is thought to have a use in the treatment for Huntington's and Parkinson's disease as it possibly corrects defects in autophagy seen in these diseases


Formula: C₁₂H₂₂O₁₁·2H₂O
MW: 378.33 g/mol
Melting Pt: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00071594
CAS Number: 6138-23-4
EINECS: 202-739-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af741ee934bfe7c82395d31c21e2c19f0738d0f0&vid=af741ee934bfe7c82395d31c21e2c19f0738d0f0&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty