Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-Tetrahydrofurfuryl alcohol ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

T2044-2.5KG T2044-500GM T2044-100GM T2044-200LTB
700008-290EA 336.45 USD InStock
700008-290 700008-294 700008-288 700008-292
(±)-Tetrahydrofurfuryl alcohol ≥99%
(±)-Tetrahydrofurfuryl alcohol
Tetrahydrofurfuryl Alcohol is used as organic synthetic reagent and a solvent for vinyl resins; dyes for leather and chlorinated rubber.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₅H₁₀O₂
Boiling Pt: 178 °C (1013 hPa)
Melting Pt: < –80 °C
Density: 1.0543 g/cm³ (20 °C)
Flash Pt: 75 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00005372
CAS Number: 97-99-4
EINECS: 202-625-6
Merck Index: 12,09353
Order Now
Specification Test Results
Assay 99% Min.

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty