Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium toluene-4-sulfonate ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP

T2182-100GM T2182-500GM T2182-2.5KG
700002-564EA 102.97 USD InStock
700002-564 700002-568 700002-566
Sodium toluene-4-sulfonate ≥99%
Sodium toluene-4-sulphonate


Formula: C₇H₇NaO₃S
Storage Temperature: Ambient
MDL Number: MFCD00798566
CAS Number: 657-84-1
EINECS: 211-522-5
Order Now
Specification Test Results
Assay 99% Min.

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty