Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Iron(II) iodide, ultra dry ≥99.99% (metals basis)

Supplier: Origen Biomedical, Inc
Danger

013619-09 013619-03
AA13619-09EA 382.58 USD InStock
AA13619-09 AA13619-03
Iron(II) iodide, ultra dry ≥99.99% (metals basis)
Iron(II) iodide
Purity: 99.99% CAS No.: 7783-86-0 Molecular Formula: FeI2 Formula Weight: 309.66 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 5.32 Appearance: Black Odor: Odorless UN No.: 3260 MDL No.: MFCD00016094


Formula: FeI₂
MW: 309.66 g/mol
Density: 5.32
Storage Temperature: Ambient
MDL Number: MFCD00016094
CAS Number: 7783-86-0
EINECS: 232-031-2
UN: 3260
ADR: 8 III
Merck Index: 12,04097
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty