Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dipraseodymium trisulphate octahydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Origen Biomedical, Inc
Warning

014607-14 014607-22
AA14607-14EA 46.5 USD InStock
AA14607-14 AA14607-22
Dipraseodymium trisulphate octahydrate ≥99.9% (REO, rare earth oxide basis)
Dipraseodymium trisulphate octahydrate
Purity: 99.9% CAS No.: 13510-41-3 Molecular Formula: Pr2(SO4)3.8H2O Formula Weight: 714.12 (570.00anhy) Physical Form: Crystalline Appearance: Green Odor: Odorless Merck Reference: 14,7712 MDL No.: MFCD00149832


Formula: O₁₂Pr₂S₃·8H₂O
MW: 714.13
Storage Temperature: Ambient
MDL Number: MFCD00149832
CAS Number: 13510-41-3
EINECS: 233-622-8
Merck Index: 14,7712
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty