Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Titanium ≥99.2% (metals basis), foil, annealed, Thickness: 1.6 mm (0.063 in)

Supplier: Origen Biomedical, Inc
Warning

044485-GH 044485-GW
AA44485-GHEA 64.4 USD InStock
AA44485-GH AA44485-GW
Titanium ≥99.2% (metals basis), foil, annealed, Thickness: 1.6 mm (0.063 in)
Titanium
Purity: 99.2% CAS No.: 7440-32-6 Physical Form: ≈72g/100x100mm Odor: Odorless MDL No.: MFCD00011264


Formula: Ti
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty