Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Aluminium ≥99.998% (metals basis), foil, Puratronic®, Thickness: 1.5 mm (0.06 in)

Supplier: Origen Biomedical, Inc

044492-FI 044492-HP 044492-GH
AA44492-FIEA 61.6 USD InStock
AA44492-FI AA44492-HP AA44492-GH
Aluminium ≥99.998% (metals basis), foil, Puratronic®, Thickness: 1.5 mm (0.06 in)
Aluminium
Purity: 99.998% CAS No.: 7429-90-5 Physical Form: ≈10.125g/50x50mm, Temper: as rolled Odor: Odorless MDL No.: MFCD00134029


Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty