Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lead ≥99.99% (metals basis), granules +30 mesh

Supplier: Origen Biomedical, Inc
Danger

045081-36 045081-A4 045081-A1
AA45081-36EA 62.68 USD InStock
AA45081-36 AA45081-A4 AA45081-A1
Lead ≥99.99% (metals basis), granules +30 mesh
Lead
Purity: 99.99% CAS No.: 7439-92-1 Appearance: Silver Odor: Odorless MDL No.: MFCD00134050


Formula: Pb
MW: 207.2 g/mol
Boiling Pt: 1740
Melting Pt: 327
Density: 11.3437 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS Number: 7439-92-1
EINECS: 231-100-4
Merck Index: 12,05410
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty