Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tin ≥99.9999% (metals basis), pieces, Puratronic®

Supplier: Origen Biomedical, Inc

045481-06 045481-14 045481-22
AA45481-06EA 94 USD InStock
AA45481-06 AA45481-14 AA45481-22
Tin ≥99.9999% (metals basis), pieces, Puratronic®
Tin
Purity: 99.9999+% CAS No.: 7440-31-5 Appearance: Silver-gray Odor: Odorless MDL No.: MFCD00133862


Formula: Sn
MW: 118.71 g/mol
Boiling Pt: 2270
Melting Pt: 232
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty