Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Molybdic acid ≥85% (MoO₃ basis)

Supplier: Origen Biomedical, Inc
Warning

A10537-36 A10537-0E A10537-22
AAAA10537-36EA 141.75 USD InStock
AAAA10537-36 AAAA10537-0E AAAA10537-22
Molybdic acid ≥85% (MoO₃ basis)
Molybdic acid
CAS No.: 7782-91-4 MDL No.: MFCD00036287


Formula: H₂MoO₄
Boiling Pt: ~ 300 °C (1013 hPa)
Melting Pt: 300 °C
Density: 3.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036287
CAS Number: 7782-91-4
EINECS: 231-970-5
Merck Index: 13,06262
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty