A12481-0E
A12481-36
AAAA12481-0CEA
360.15
USD
InStock
AAAA12481-0C
AAAA12481-0E
AAAA12481-36
Benzyl chloride 99% stabilized
Benzyl chloride
MDL: MFCD00000889
Beilstein: 471308
Miscible with alcohol, ether. Insoluble in water
Purity: 99%
CAS No.: 100-44-7
Molecular Formula: C7H7Cl
Formula Weight: 126.59
Flash Point: 73°(163°F)
Physical Form: Liquid
Density: 1.099
Refractive Index: 1.5380
Melting Point: -43°
Boiling Point: 178-180°
Solubility: Miscible with alcohol, ether. Insoluble in water
Beilstein Reference: 471308
Merck Reference: 14,1129
UN No.: 1738
MDL No.: MFCD00000889