Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dicyclopentadiene 90+% stabilized

Supplier: Origen Biomedical, Inc
Danger

A13643-0F A13643-AP
AAA-A13643-0FEA 97.67 USD InStock
AAA-A13643-0F AAA-A13643-AP
Dicyclopentadiene 90+% stabilized
Dicyclopentadiene


Formula: C₁₀H₁₂
Boiling Pt: 171…173 °C
Melting Pt: 12 - 34 °C
Density: 0.982 g/cm³ (20 °C)
Flash Pt: 26 °C (78 °F)
Storage Temperature: Ambient
MDL Number: MFCD00078246
CAS Number: 77-73-6
EINECS: 201-052-9
UN: 2048
ADR: 3,III
Merck Index: 13,02767
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty