Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chlorophenylhydrazinium chloride 97%

Supplier: Origen Biomedical, Inc
Warning

A14518-22 A14518-06 A14518-14
AAA14518-22EA 332 USD InStock
AAA14518-22 AAAA14518-06 AAAA14518-14
4-Chlorophenylhydrazinium chloride 97%
4-Chlorophenylhydrazinium chloride
Purity: 97% CAS No.: 1073-70-7 Molecular Formula: C6H7ClN2.HCl Formula Weight: 179.05 Odor: Odorless Melting Point: ca 216° dec. Beilstein Reference: 3596254 MDL No.: MFCD00012943


Formula: C₆H₈Cl₂N₂
Storage Temperature: Ambient
MDL Number: MFCD00012943
CAS Number: 1073-70-7
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty