A17901-06
A17901-14
A17901-22
AAAA17901-06EA
90.6
USD
InStock
AAAA17901-06
AAAA17901-14
AAAA17901-22
Phenylsilane 97%
Phenylsilane
Purity: 97%
CAS No.: 694-53-1
Molecular Formula: C6H8Si
Formula Weight: 108.22
Flash Point: 7°(45°F)
Refractive Index: 1.5105
Boiling Point: 120°
UN No.: 3399
MDL No.: MFCD00013478