Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tributyl borate 98%

Supplier: Origen Biomedical, Inc
Warning

A19322-0F A19322-AE A19322-AP
AAAA19322-0FEA 624 USD InStock
AAAA19322-0F AAAA19322-AE AAAA19322-AP
Tributyl borate 98%
Tributyl borate
Purity: 98% CAS No.: 688-74-4 Molecular Formula: [CH3(CH2)3O]3B Formula Weight: 230.16 Flash Point: 93°(199°F) Refractive Index: 1.4095 Melting Point: -70° Boiling Point: 230-235° Beilstein Reference: 1703865 MDL No.: MFCD00009434


Formula: C₁₂H₂₇BO₃
MW: 230.15 g/mol
Boiling Pt: 230…235 °C
Melting Pt: < –70 °C
Density: 0.853
Flash Pt: 93 °C(199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009434
CAS Number: 688-74-4
EINECS: 211-706-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty