B21200-14
B21200-06
AAAB21200-14EA
68.39
USD
InStock
AAAB21200-14
AAAB21200-06
4-Chloro-3-(trifluoromethyl)aniline 99%
4-Chloro-3-(trifluoromethyl)aniline
Purity: 99%
CAS No.: 320-51-4
Molecular Formula: C7H5ClF3N
Formula Weight: 195.57
Flash Point: 134°(273°F)
Melting Point: 35-38°
Boiling Point: 85°/3mm
Beilstein Reference: 641588
UN No.: 2811
MDL No.: MFCD00007846