B22131-0E
B22131-36
AAB22131-0EEA
134
USD
InStock
AAB22131-0E
AAB22131-36
2-Anilinoethanol 98%
2-Anilinoethanol
Purity: 98%
CAS No.: 122-98-5
Molecular Formula: C6H5NH(CH2)2OH
Formula Weight: 137.18
Flash Point: 153°(307°F)
Refractive Index: 1.5790
Odor: Amine-like
pH: 10.2
Melting Point: -30°
Boiling Point: 284-286°
Beilstein Reference: 774672
UN No.: 2810
MDL No.: MFCD00002832