Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dichloroacetyl chloride 97%

Supplier: Origen Biomedical, Inc
Danger

B22296-22 B22296-36
AAB22296-22EA 46.24 USD InStock
AAB22296-22 AAB22296-36
Dichloroacetyl chloride 97%
Dichloroacetyl chloride
Purity: 97% CAS No.: 79-36-7 Molecular Formula: Cl2CHCOCl Formula Weight: 147.39 Flash Point: None Refractive Index: 1.4620 Boiling Point: 107-108° Merck Reference: 14,3053 UN No.: 1765 MDL No.: MFCD00000840


Formula: C₂HCl₃O
MW: 147.39 g/mol
Boiling Pt: 107
Density: 1.532
Flash Pt: 66
Storage Temperature: Ambient
MDL Number: MFCD00000840
CAS Number: 79-36-7
EINECS: 201-199-9
UN: 1765
ADR: 8 II
Merck Index: 14,3053
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty