Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Anisoyl chloride 97%

Supplier: Origen Biomedical, Inc
Danger

B22784-14 B22784-06 B22784-22
AAB22784-14EA 134 USD InStock
AAB22784-14 AAB22784-06 AAB22784-22
2-Anisoyl chloride 97%
2-Anisoyl chloride
Purity: 97% CAS No.: 21615-34-9 Molecular Formula: C8H7ClO2 Formula Weight: 170.60 Flash Point: 84°(184°F) Refractive Index: 1.5724 Boiling Point: 128-129°/8mm Beilstein Reference: 607605 UN No.: 1729 MDL No.: MFCD00000664


Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Boiling Pt: 137
Density: 1.146
Flash Pt: 84 °C (184 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000664
CAS Number: 21615-34-9
EINECS: 244-477-5
UN: 1729
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty