Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

β-Propiolactone 97%

Supplier: Origen Biomedical, Inc
Danger

B23197-06 B23197-14 B23197-03
AAB23197-06EA 267 USD InStock
AAB23197-06 AAB23197-14 AAB23197-03
β-Propiolactone 97%
β-Propiolactone
Purity: 97% CAS No.: 57-57-8 Molecular Formula: C3H4O2 Formula Weight: 72.06 Storage Temperature: -30°C to -10°C Flash Point: 70°(158°F) Physical Form: Liquid Density: 1.146 Refractive Index: 1.4130 Sensitivity: Moisture Sensitive Appearance: Colorless Melting Point: -34° Boiling Point: 161-163° Merck Reference: 14,7820 UN No.: 3382 MDL No.: MFCD00005169


Formula: C₃H₄O₂
MW: 72.06 g/mol
Boiling Pt: 155
Melting Pt: -31.2
Density: 1.146
Flash Pt: 70 °C (158 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00005169
CAS Number: 57-57-8
EINECS: 200-340-1
UN: 3382
ADR: 6.1 I
Merck Index: 14,7820
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty