B23928-06
B23928-22
B23928-14
AAB23928-06EA
33.7
USD
InStock
AAB23928-06
AAB23928-22
AAB23928-14
Pentafluoroanisole 98%
Pentafluoroanisole
Purity: 98%
CAS No.: 389-40-2
Molecular Formula: C7H3F5O
Formula Weight: 198.09
Flash Point: 32°(89°F)
Refractive Index: 1.4090
Melting Point: -38°
Boiling Point: 136-138°
Beilstein Reference: 2052664
UN No.: 1993
MDL No.: MFCD00000289