Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Perhydroazepine 98+%

Supplier: Origen Biomedical, Inc
Danger

B24007-AE B24007-AP
AAB24007-AEEA 27.4 USD InStock
AAB24007-AE AAB24007-AP
Perhydroazepine 98+%
Perhydroazepine
Purity: 98+% CAS No.: 111-49-9 Molecular Formula: C6H13N Formula Weight: 99.18 Flash Point: 22°(71°F) Refractive Index: 1.4660 Odor: Ammonia-like Melting Point: -37° Boiling Point: 138-139° Beilstein Reference: 1084 UN No.: 2493 MDL No.: MFCD00006934


Formula: C₆H₁₃N
MW: 99.18 g/mol
Boiling Pt: 138
Melting Pt: -37
Density: 0.880
Flash Pt: 22 °C (71 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006934
CAS Number: 111-49-9
EINECS: 203-875-9
UN: 2493
ADR: 3 II
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty